Inlet Filter, Labconco® | Avantor
Inlet Filter, Labconco®
Supplier:   Labconco
Inlet Filter, Labconco®
Supplier:  Labconco
Certificates
Inlet Filter, Labconco®
Inlet Filter, Labconco®
2 Options Available Below
Product Details & Documents
This disposable filter prevents oil backstreaming from the vacuum pump and protects the pump from damage.
Filter life is approximately 1000 hours at 10 µm vacuum.
Delivery information: Includes filter assembly and one filter cartridge.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Inlet Filter
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,280.00
    $1,280.00
    Each
       $1,280.00
    La quantité minimale de commande est de 1
    Supplier Number: 1472200
  • Replacement Filter Cartridge
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,100.00
    $1,100.00
    Each
       $1,100.00
    La quantité minimale de commande est de 1
    Supplier Number: 1472500