Centering Ring Assemblies, Welch® (Gardner Denver)
Centering Ring Assemblies, Welch® (Gardner Denver)
Supplier:   Welch Vacuum
Centering Ring Assemblies, Welch® (Gardner Denver)
Supplier:  Welch Vacuum
Certificates
Centering Ring Assemblies, Welch® (Gardner Denver)
Centering Ring Assemblies, Welch® (Gardner Denver)
2 Options Available Below
Product Details & Documents
Stainless steel centering ring with replaceable Viton® O-ring. Use to 150 °C sustained, 200 °C intermittent.
Product Family Options
Product Information
  • Ø int.
    ISO Size
    Availability
    Price
  • 40 mm (19/16")
    NW40
    Log in for availability
    Log in for availability
    Each
    Each - $86.06
    $86.06
    Each
       $86.06
    Supplier Number: 303103
  • 16 mm (5/8")
    NW16
    Log in for availability
    Log in for availability
    Each
    Each - $58.70
    $58.70
    Each
       $58.70
    Supplier Number: 303101