ContactUsLinkComponent
Digital 'Pocket' Urine Specific Gravity Refractometer, ATAGO®
Catalog # 80087-520
Supplier:  Atago
undefined
Digital 'Pocket' Urine Specific Gravity Refractometer, ATAGO®
Catalog # 80087-520
Supplier:  Atago
Supplier Number:  4410
Specifications
  • Accuracy:
    ±0.001 specific gravity
  • Measurement range:
    1.0000 - 1.0600 specific gravity
  • Resolution:
    0.0001 specific gravity
  • Temperature:
    10...35 °C (50...95 °F) measurement and ambient
  • Weight:
    100 g (3.5 oz.)
  • Cat. no.:
    80087-520
  • W×D×H:
    5.5×3.1×10.9 cm (23/16×11/8×45/16")
  • Supplier No.:
    4410
Specifications
About this item
Digital handheld refractometer provides an instant LCD reading of urine specific gravity from a small sample of urine placed on the prism
The unit is simple to operate, dust-tight, and protected against water jets. It features automatic temperature compensation and IP65 protection. Uses two AAA batteries.