PerfeCTa® MultiPlex qPCR SuperMix
PerfeCTa® MultiPlex qPCR SuperMix
 :  Quantabio
PerfeCTa® MultiPlex qPCR SuperMix
PerfeCTa® MultiPlex qPCR SuperMix
This SuperMix is a 2x concentrated, ready-to-use reaction cocktail for real-time quantitative PCR (qPCR) that contains all components, except primers, probes, and templates
  • Ultrapure, hot start AccuStart enzyme technology and anti-foaming technology
  • Available in two versions: Universal and Low-ROX
  • Maximum assay sensitivity and target precision with stringent, ultrapure, and high-yielding AccuStart II hot start Taq DNA polymerase
  • Broad linear detection range with cDNA or genomic DNA templates
  • Enables highly sensitive 4 target detection assays with coamplified, robust internal positive controls
  • Supports efficient vortex mixing with proprietary anti-foaming technology
A key component of this supermix is AccuStart™ Taq DNA polymerase with monoclonal antibodies that bind to the polymerase and keep it inactive prior to the initial PCR denaturation step. Upon heat activation (2 minutes at 95°C), the antibodies denature irreversibly, releasing fully active, unmodified Taq DNA polymerase. This enables specific and efficient primer extension with the convenience of room temperature reaction assembly.
The system transcends multiplex limitations of conventional PCR master mixes, enabling unbiased amplification of up to five target sequences in a single tube. Suppression of low copy amplicons by high copy reference targets in the amplification is a common problem in multiplex PCR. This can skew, or mask the apparent representation and quantification of low copy target sequences. PerfeCTa® MultiPlex qPCR SuperMix delivers dynamic range and sensitivity to multiplexed qPCR that is comparable to that for singleplex qPCR probe assays without the need for limiting or variable primer concentrations.
  • Catalog No:
  • CA101414-204
  • CA89168-786
  • CA97065-230
  • 97065-230
  • CA97065-232
  • Application:
  • qPCR probe
  • qPCR probe
  • qPCR probe
  • qPCR probe
  • qPCR probe
  • Multiplexing:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
Product Family Options
  • Désignation
    Compatibilité du cycleur
    No. of Reactions
  • PerfeCTa® MultiPlex qPCR SuperMix
    Agilent AriaMx, BioRad CFX, QIAGEN Rotor-Gene Q, Quantabio Q, Roche LightCycler 480
    200× 50 µl
    Each
    Each - $1,055.33
    $1,055.33
    Each
       $1,055.33
     : 95063-200
    Specifications
    Cat. No.
    CA101414-204
    Application
    qPCR probe
    Multiplexing
    Yes
  • PerfeCTa® MultiPlex qPCR SuperMix
    Agilent AriaMx, BioRad CFX, QIAGEN Rotor-Gene Q, Quantabio Q, Roche LightCycler 480
    1000× 50 µl
    Each
    Each - $5,238.77
    $5,238.77
    Each
       $5,238.77
     : 95063-01K
    Specifications
    Cat. No.
    CA89168-786
    Application
    qPCR probe
    Multiplexing
    Yes
  • PerfeCTa® MultiPlex qPCR SuperMix, Low ROX™
    ABI 7500/7500 Fast/QuantStudio™, Agilent AriaMx, Douglas Scientific IntelliQube®, Stratagene Mx
    200× 50 µl
    Each
    Each - $832.41
    $832.41
    Each
       $832.41
     : 95108-200
    Specifications
    Cat. No.
    CA97065-230
    Application
    qPCR probe
    Multiplexing
    Yes
  • PerfeCTa® MultiPlex qPCR SuperMix, Low ROX™
    ABI 7500/7500 Fast/QuantStudio™, Agilent AriaMx, Douglas Scientific IntelliQube®, Stratagene Mx
    200× 50 µl
     : 95108-200
    Specifications
    Cat. No.
    97065-230
    Application
    qPCR probe
    Multiplexing
    Yes
  • PerfeCTa® MultiPlex qPCR SuperMix, Low ROX™
    ABI 7500/7500 Fast/QuantStudio™, Agilent AriaMx, Douglas Scientific IntelliQube®, Stratagene Mx
    1000× 50 µl
    Each
    Each - $4,028.92
    $4,028.92
    Each
       $4,028.92
     : 95108-01K
    Specifications
    Cat. No.
    CA97065-232
    Application
    qPCR probe
    Multiplexing
    Yes