About this item
FastMix® reaction mixes include buffers and stabilizers optimized for maximum PCR efficiency, reproducibility, and sensitivity with TaqMan® and TaqMan® MGB probe chemistries
- Fast Cycling protocols
- Contains UNG to eliminate amplification of carry-over contamination
Kits are available for all real-time PCR instrument platforms including those requiring normalization with ROX™ reference dye. Kits 101419-202, -204, and -206 are for use with platforms not requiring passive reference dye, including Roche LightCycler® 480, Bio-Rad® iQ™, MyiQ™, iQ™5, Opticon®, and Chromo4™, Corbett Rotor-Gene™, Eppendorf® Mastercycler®, and Cepheid® SmartCycler®. Kits with Low ROX™ reference dye are for use with platforms including AB 7500 and Stratagene® Mx™. Kits with ROX™ reference dye are for use with platforms including AB 7000, 7300, 7700, 7900, and StepOne™.
A key component of PerfeCTa® qPCR FastMix® reaction mixes is AccuFast™ Taq DNA polymerase that enables instant activation and rapid polymerization kinetics. Upon heat activation (20 seconds at 95°C), the antibodies irreversibly denature releasing fully active, unmodified Taq DNA polymerase, which promotes specific and efficient primer extension with the convenience of room temperature reaction assembly. FastMix® reaction mixes deliver maximum PCR efficiency, sensitivity, specificity, and a robust fluorescent signal with either fast or conventional cycling conditions.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Application:qPCR probe