Mini Iris Scissors | Avantor
Mini Iris Scissors
Mini Iris Scissors
Certificates
Mini Iris Scissors
1 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Iris scissors were originally designed for ophthalmic procedures but are now used in a wide range of applications. The tips are sharp, and they come in curved, straight or angled styles.
Product Family Options
Informations sur le produit
  • Désignation
    Longueur
    Matière
    Curvature
    Availability
    Price
  • Mini iris scissor, sharp tips
    8 cm (3.1")
    Stainless steel
    Curved
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $82.73
    $82.73
    Each
       $82.73
    Supplier Number: 503671
  • Mini iris scissor, sharp tips
    8 cm (3.1")
    Stainless steel
    Straight
    Discontinued
    Supplier Number: 503670