Mini Dissecting Scissors, Left Hand
Mini Dissecting Scissors, Left Hand
Certificates
Mini Dissecting Scissors, Left Hand
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Our Left-Hand Mini Dissecting Scissors are similar to regular dissecting scissors, but they are shorter and have more delicate tips, making them ideal for fine dissection work in tight spaces.
  • Our dissecting scissors are for general use in dissection, preparation of tissue samples, and small animal surgeries
  • Have thinner (and often longer) blades than operating scissors
  • Designed for cutting and probing in tight spaces
  • Designed for left handed people
They are commonly used in microsurgery, like ophthalmic surgery. These scissors are 9 cm (3.5") long and have straight blades with sharp tips or curved blades with fine tips.
These are available in surgical grade stainless steel. Our Dissecting Scissors are designed to withstand the high heat and chemical exposure of the sterilization process. They may be autoclaved. To guarantee a long life for your surgical instruments, be sure to clean them thoroughly after each use.
Product Family Options
Informations sur le produit
  • Désignation
    Longueur
    Matière
    Curvature
    Availability
    Price
  • Mini dissecting scissor, fine tips
    9 cm (3.5")
    Stainless steel
    Curved
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $99.78
    $99.78
    Each
       $99.78
    Supplier Number: 503627
    Spécifications:
    More
    Specifications
    Cat. No.
    77663-338
  • Mini dissecting scissor, sharp tips
    9 cm (3.5")
    Stainless steel
    Straight
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $99.78
    $99.78
    Each
       $99.78
    Supplier Number: 503625
    Spécifications:
    More
    Specifications
    Cat. No.
    77663-336