Blade for Integraslice | Avantor
Blade for Integraslice
Blade for Integraslice
Certificates
Blade for Integraslice
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
For use with integraslice vibrating microtome CA-5100MZ and CA-5100MZ Plus.
Product Family Options
Informations sur le produit
  • Désignation
    Matière
    Availability
    Price
  • Microtome blade, pack of 5
    Ceramic
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,861.24
    $1,861.24
    Each
       $1,861.24
    Supplier Number: 501631
  • Microtome blade, pack of 50
    Stainless steel
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $842.00
    $842.00
    Each
       $842.00
    Supplier Number: 501632