Carbon Wire | Avantor
Carbon Wire
Carbon wire, 30 µm (0.0012"), 1.5 m (5')
Catalog # 77664-078
Fournisseur #:  C3005
Carbon Wire
Catalog # 77664-078
Carbon Wire
Carbon wire, 30 µm (0.0012"), 1.5 m (5')
Price
$142.22
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Carbon wire is a single 30 µm fiber of electrochemically activated carbon. This fiber is especially useful in microelectrochemical experiments.
  • 49 AWG
  • 0.0012" (30 µm) Ø
  • 5' (1.5 m) long
Spécifications
  • Ø:
    30 µm (0.0012")
  • Désignation:
    Carbon wire
  • Longueur:
    1.5 m (5')