Universal Dipping Probe Stand
The universal dipping probe stand is designed to holds dip tips, diptip minis, spectropipetters and biosensors (macro and micro).
: Kit includes: Small V-base, 25 cm stainless steel post and holder bracket, delrin with thumbscrew
- Désignation:Universal dipping probe stand