Flexible Light Guides
Flexible Light Guides
Flexible Light Guides
Certificates
Flexible Light Guides
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Single pipe light guide with focusing lens.
Standard adapter is available for attaching to a fiber optic illuminator like the Z-Lite series.
Fiber diameter: 6 mm
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Light guide, 18" (460 mm) long, ⁵/₈" standard adapter
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $486.59
    $486.59
    Each
       $486.59
    Supplier Number: 504930
  • Light guide, 24" (610 mm) long, ⁵/₈" standard adapter
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $802.91
    $802.91
    Each
       $802.91
    Supplier Number: 504931