VWR® 901 Portable Dissolved Oxygen Meter (Optical)
77618-940
New Product
- Description:VWR-DO901 meter with DO3 optical DO electrode and zero oxygen calibration solution powder vial
- Cat. no.:77618-940
The VWR-DO901 meter provide accurate and reliable measurements of dissolved oxygen concentration and dissolved oxygen saturation with an optical DO electrode.
- High resolution color touch display (4.3")
- Air-saturated water or zero oxygen calibration capabilities
- IP 65 waterproof
- Automatic or manual temperature compensation capabilities
- Auto-read, timed-read, and continuous-read measurement options
- Data capacity of 1000 sets for each parameter (GLP-compliant)
- Rechargeable battery
- Auto barometric pressure compensation
The easy to use VWR-DO901 portable meters provide accurate dissolved oxygen measurements for accurate water quality testing. The intelligent operation system provides features including user management, calibration management, electrode management, method management, and many other advanced capabilities.
: For use with DO3 optical DO electrode. Three-year meter warranty. For application specific recommendations, please reference 'VWR® Electrode Selection Guide' located in the Related Information section.
: Electrode included. Meter also includes electrode holders (2), meter silicone rubber case, carrying case, touch screen pen and hand strap.