Mouse Rectal Probe Ball Tip | Avantor
Mouse Rectal Probe Ball Tip
Mouse rectal probe ball tip, 1.1 m
Catalog # 77663-918
Fournisseur #:  61824
Mouse Rectal Probe Ball Tip
Catalog # 77663-918
Mouse Rectal Probe Ball Tip
Mouse rectal probe ball tip, 1.1 m
Price
$622.52
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The needle microprobes and animal rectal probes are all designed to withstand sterilization by ETO gas and gluteraldehyde solution. Our flexible implantable microprobes are both ETO gas sterilizable and autoclavable.
  • Rectal probe for use with the ATC2000, Animal temperature controller
  • The heating plate cable is 1.1 m long
Spécifications
  • Désignation:
    Mouse rectal probe ball tip
  • Longueur:
    1.1 m