Sterilization Mesh Cassettes | Avantor
Sterilization Mesh Cassettes
Sterilization Mesh Cassettes
Certificates
Sterilization Mesh Cassettes
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Mesh sterilization baskets are perfect for sterilizing, storing and organizing small components or instrument. These baskets are manufactured from 304 stainlesss steel. They are non-magnetic and electropolished.
  • These baskets come with a wire locking clip
  • This style of sterilization baskets is available in three sizes.
They are burr-free to protect the instrument wraps from tearing. The fine mesh has open squares of 1.6 mm, and the wire edge diameter is 0.7 mm.
Product Family Options
Informations sur le produit
  • Dimensions
    Availability
    Price
  • 4×4×2 cm (1.5×1.5×.75")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $90.03
    $90.03
    Each
       $90.03
    Supplier Number: 504766
  • 8×8×3.4 cm (3.1×3.1×1.4")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $99.78
    $99.78
    Each
       $99.78
    Supplier Number: 504767
  • 10.5×7×2.5 cm (4.1×2.75×1")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $138.69
    $138.69
    Each
       $138.69
    Supplier Number: 504768