Stereotaxic Kit | Avantor
Stereotaxic Kit
Sterotaxic surgical kit
Catalog # 77663-760
Fournisseur #:  504743
Stereotaxic Kit
Catalog # 77663-760
Stereotaxic Kit
Sterotaxic surgical kit
Price
$469.59
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Four piece sterotaxic surgical kit including popular instruments.
Delivery information: Kit includes: Scalpel handle #3, 13 cm, Metzenbaum scissors, 11.5 cm, SuperCut, straight, microdissecting forceps, 10.2 cm, angled, serrated, needle holder, 14 cm, round handles, straight.
Spécifications
  • Désignation:
    Sterotaxic surgical kit