MICRO-ePORE™ Electrode Holders
MICRO-ePORE™ Electrode Holders
MICRO-ePORE™ Electrode Holders
These electrode holders are designed exclusively for use the with MICRO-ePORE™ system, which includes the silver wire electrode holders. The platinum wire electrode holders are an optional accessory.
Platinum wire electrode holders are important when you are working with proteins. The silver chloride wire is perfect for work with RNA. However, the silver chloride wire tends to denature proteins, reducing the efficiency of the microinjection solution.
Applications for silver wire electrode holders: Silver wire electrodes are suitable for most applications
Applications for platinum wire electrode holders: Platinum electrodes offers more active insertion of the genetic material capability, and there is a better chance of changing the genome
Product Family Options
  • For Use With
    Includes
    Désignation
  • WPI MICRO-ePORE pinpoint cell penetrator when using a WPI PV850 or uPUMP injector
    PicoNozzle tip assemblies (Handle diameter is 6.25 mm and 100 mm long, 5' tubing, 1.0 mm pipette gaskets (lime green), 1.2 mm pipette gaskets (black), 1.5 mm pipette gaskets (red), 1.65 mm pipette gaskets (white), sealing gaskets
    WPI MICRO-ePORE holder, platinum wire
    Each
    Each - $556.16
    $556.16
    Each
       $556.16
     : 300683PT
  • WPI MICRO-ePORE pinpoint cell penetrator when using a Eppendorf Femtojet® injector
    1.0 mm pipette gaskets (lime green), 1.2 mm pipette gaskets (black), 1.5 mm pipette gaskets (red), 1.65 mm pipette gaskets (white), sealing gaskets
    Femtojet MICRO-ePORE holder, silver wire
    Each
    Each - $896.05
    $896.05
    Each
       $896.05
     : 300684
  • WPI MICRO-ePORE pinpoint cell penetrator when using a Eppendorf Femtojet® injector
    1.0 mm pipette gaskets (lime green), 1.2 mm pipette gaskets (black), 1.5 mm pipette gaskets (red), 1.65 mm pipette gaskets (white), sealing gaskets
    Femtojet MICRO-ePORE holder, platinum wire
    Each
    Each - $556.16
    $556.16
    Each
       $556.16
     : 300684PT
  • WPI MICRO-ePORE pinpoint cell penetrator when using a Narishige injector
    1.0 mm pipette gaskets (lime green), 1.2 mm pipette gaskets (black), 1.5 mm pipette gaskets (red), 1.65 mm pipette gaskets (white), sealing gaskets
    Narishige MICRO-ePORE holder, silver wire
    Each
    Each - $877.53
    $877.53
    Each
       $877.53
     : 300685