Catherization Cannulation Kit
Catalog # 77663-758
Supplier: WORLD PRECISION INSTRUMENTS LLC
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Catherization and cannulation kit
- Material:Stainless steel
- Cat. no.:77663-758
Specifications
About this item
Nine piece surgical instrument kit ideal for catherization and cannulation.
Delivery: Kit includes: Disposable scalpel, #15, sterile, 10/box, lever action skin stapler 35S, sterile, Metzenbaum scissors, 11.5 cm, SuperCut, straight, castroviejo needle holder with lock, 14 cm, straight, serrated, Halsted mosquito hemostatic forceps, 12.5 cm, straight, Spence stitch ligature scissors, 9 cm, Adson-Brown forceps, 12 cm, straight, TC jaws, 7×7 teeth, WPI Swiss tweezers #5, 11 cm, 0.1×0.06 mm tips, Nugent utility forceps, 10 mm, angled tip.