Microscope, Binocular with Plan Achromat LED | Avantor
Microscope, Binocular with Plan Achromat LED
Binocular microscope, with plan achromat LED
Catalog # 77663-564
Fournisseur #:  504220
Microscope, Binocular with Plan Achromat LED
Catalog # 77663-564
Microscope, Binocular with Plan Achromat LED
Binocular microscope, with plan achromat LED
Price
$4,814.15
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
ACCU-SCOPE's 3012 Series features E-Plan Achromat Infinity objectives which are renowned for crisp, high contrast images with outstanding resolution. Modular in design, the 3012 boasts options for entry level fluorescence applications, phase contrast, darkfield and simple polarization.
  • Binocular
  • Plan achromat optics
With 22 mm field of view eyepieces and an eyepoint height that is adjustable, it can be raised 35 mm by rotating the eyepiece tubes up 180° to accommodate the needs of various users
ACCU-SCOPE's Infinity E-plan objectives provide users with best in class high resolution, high contrast objectives. Objectives are available in plan achromat and plan phase configurations.
The rackless, scratch resistant rectangular stage has low-positioned X-Y coaxial controls. A clinical level microscope for routine applications in medical, veterinary and university labs.
Spécifications
  • Désignation:
    Binocular microscope, with plan achromat LED