ContactUsLinkComponent
Quantrex® Ultrasonic Cleaner
  77663-560
New Product
Quantrex® Ultrasonic Cleaner
  77663-560
 :  504216
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Ultrasonic cleaner
  • Timer:
    60 mintues
  • Cat. no.:
    77663-560
Quantrex® bath with a timer.
  • Provides super-strength cleaning every time
  • When used with L&R’s specialty formulated solutions, the self-contained, compact unit offers efficient trouble-free cleaning
  • Each Quantrex machine comes standard with increased power—strength you can see as soon as you turn the unit on
  • Stainless steel drain with multi-positional outlet for easy removal of solution
  • 14 quality inspection steps for strength and durability
It is made of vinyl-stainless steel and stainless steel.
Application: Versatile enough for a variety of cleaning applications.
 : Includes timer, drain and cover and heater.