LED Ring Light | Avantor
LED Ring Light
LED ring light, US plug, 90 - 264 VAC, 50/60 Hz
Catalog # 77663-500
Fournisseur #:  504134
LED Ring Light
Catalog # 77663-500
LED Ring Light
LED ring light, US plug, 90 - 264 VAC, 50/60 Hz
Price
$673.95
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
'White' light illumination with 72 LED bulbs.
  • Maximum opening 61 mm
  • Brightness adjustable
  • ESD safe
  • Direct top illumination plus four zones for shadows
  • Low cost
  • Less bulky than halogen
  • Cold light with no heat
Ring light is divided into four areas and each area is turned on and off separately.
Application: Stereo microscope use
Spécifications
  • Désignation:
    LED ring light, US plug
  • Electrical:
    90 - 264 VAC, 50/60 Hz