WPI Vannas Spring Scissors
WPI Vannas Spring Scissors
WPI Vannas Spring Scissors
Our Vannas scissors are specialized spring scissors designed for precision cutting and often used by scientific researchers. They are named after the Swedish scientist Gunnar Vannas who invented them in the 1940s.
  • 0.025×0.015 mm tips
  • 7 mm Super fine blades
Compared with other spring scissors, Vannas scissors are distinguished by their delicate, thin, sharp tips. These surgical scissors have a super fine blade and 0.025×0.015 mm fine tips. The slender, pointed tips allows for precise cutting with minimal damage to surrounding tissues. Choose between straight or curved blades.
Manufactured with surgical grade stainless steel, these scissors are durable and resistant to corrosion. As with all your surgical instruments, these scissors should be cleaned immediately after use to ensure a long service life and minimize staining or corrosion. These Vannas are autoclavable.
Product Family Options
  • Désignation
    Longueur
    Matière
  • WPI vannas spring scissors, super fine, straight
    8.5 cm (3")
    Stainless steel
    Each
    Each - $688.56
    $688.56
    Each
       $688.56
     : 500086
  • WPI vannas spring scissors, super fine, curved
    8.5 cm (3")
    Stainless steel
    Each
    Each - $664.22
    $664.22
    Each
       $664.22
     : 501232