Gillies Dissecting Forceps
Gillies Dissecting Forceps
Gillies Dissecting Forceps
Gillies Dissecting Forceps
Gillies Dissecting Forceps, also known as Gillies forceps or Gillies skin hooks, are often used in plastic and reconstructive surgery, and are perfect for the fine work of microdissection. The tips are narrow, pointed, and serrated, allowing you deeper access into incisions. The serrated and toothed tips, providing a secure grip on delicate tissues without causing excessive trauma or damage.
  • 15.5 cm long
  • Straight, serrated tips
  • 1×2 teeth
  • Stabilizing pin
  • Textured handle for better control
  • Plain or Tugsten carbide inserts
  • Surgical grade stainless steel or premium German steel
The narrow design of the Gillies forceps tips allow for precise dissection and manipulation of small structures, such as blood vessels, nerves, and delicate skin flaps. They come as standard or with tungsten carbide inserts. Tungsten carbide (TC) is extremely hard and durable, giving you a long-lasting pair of forceps. With a high coefficient of friction, TC forceps offer enhanced gripping ability for a secure hold with minimal friction against tissues.
The reduced friction means less tissue damage as a result of handling. Tungsten carbide is also resistant to corrosion. Our Gillies Dissecting Forceps are manufactured with surgical grade stainless steel or premium German steel, both of which are autoclavable. WPI recommends properly cleaning all your surgical instruments promptly after use. This improves the useable lifespan of your surgical tools.
Product Family Options
  • Désignation
    Matière
    Longueur
    Tip Configuration
  • Gillies dissecting forcep
    Stainless steel
    15.5 cm
    1×2 teeth
    Each
    Each - 189,81 $
    189,81 $
    Each
       189,81 $
     : 501265
  • Gillies dissecting forcep, german
    Stainless steel
    15.5 cm
    1×2 teeth
    Each
    Each - 191,18 $
    191,18 $
    Each
       191,18 $
     : 501265-G
  • Gillies dissecting forcep
    Stainless steel
    15.5 cm
    1×2 teeth
    Each
    Each - 124,09 $
    124,09 $
    Each
       124,09 $
     : 501266
  • Gillies dissecting forcep, german
    Stainless steel
    15.5 cm
    1×2 teeth
    Each
    Each - 116,92 $
    116,92 $
    Each
       116,92 $
     : 501266-G