Dissecting Needles
Dissecting Needles
Dissecting Needles
Certificates
Dissecting Needles
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
A dissecting needle, also known as a dissecting probe or dissecting pin, is used in dissection procedures to manipulate and separate tissues, examine specimens, or perform precise dissection techniques.
  • One time use
  • Stainless steel
  • Sharp
Our dissecting needle is 14 cm (5.5") long and has a plastic handle for a solid and comfortable grip. The thin, pointed metallic shaft is available as straight or angled. Dissecting needles are used to gently separate and lift tissues during dissection, or for holding or positioning small specimens or structures. With their fine and sharp tips, dissecting needles are also handy for microdissection procedures involving microscopic structures.
The shaft is stainless steel, but these are not autoclavable. They are designed to be disposable. Each package contains 12 dissecting needles.
Product Family Options
Informations sur le produit
  • Désignation
    Matière
    Longueur
    Packed
    Availability
    Price
  • Dissecting needle, straight
    Stainless steel
    14 cm (5.5")
    12/pack
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 12
    Pack of 12 - $38.96
    $38.96
    Pack of 12
       $38.96
    Supplier Number: 500458
  • Dissecting needle, angled
    Stainless steel
    14 cm (5.5")
    12/pack
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 12
    Pack of 12 - $41.36
    $41.36
    Pack of 12
       $41.36
    Supplier Number: 500460