Scissors, Biemer | Avantor
Scissors, Biemer
Scissors, biemer, tip Ø 2 mm, 13 cm (5"), Stainless steel
Catalog # 77662-340
Fournisseur #:  500381
Scissors, Biemer
Catalog # 77662-340
Scissors, Biemer
Scissors, biemer, tip Ø 2 mm, 13 cm (5"), Stainless steel
Price
$255.47
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Cuts vessels perpendicularly and without slipping.
  • Spring handle
  • Straight
  • Stainless steel
Spécifications
  • Désignation:
    Scissors, biemer, tip Ø 2 mm
  • Longueur:
    13 cm (5")
  • Matière:
    Stainless steel