Metallurgy Objective
Objective, 40×, 0.25
  77663-976
Metallurgy Objective
  77663-976
Metallurgy Objective
Objective, 40×, 0.25
$1,044.81
Each
  • Long working distance objective for a microscope
  • 40× Magnification
  • 0.25 Numerical aperature
  • Grossissement:
    40×
  • Désignation:
    Objective
  • Aperture:
    0.25