Fixing Screws
Product Details & Documents
Fixing screw for securing the internal cannula to the guide cannula, to prevent movement during injection.
- One size fits all
Specifications
- Désignation:Fixing screw for connecting injector and guide cannula