Axis Adjustment Tool
Product Details & Documents
Micromanipulator adjustment tool to modify the anti-drift tension.
- Refer to the manual for instructions on use
- For use with the KITE and M3301 micromanipulators
Spécifications
- Désignation:Micromanipulator adjustment tool
- For Use With:KITE and M3301 micromanipulators