À propos de cet article
The titanium scalpel handle is for use with our Sapphire blades.
- Titanium is non-magnetic, non-conductive, non-corrosive
- Retractable
- Reusable
Since sapphires are one of the hardest materials available (second only to diamonds), they make super sharp blades for precise, clean cuts. They are non-magnetic, especially when combined with a titanium handle. Sapphire blades are non-conductive, which may be important if you are using electrical instruments.
Sapphire blades are impervious to corrosion and extremely durable. These handles are retractable, so you can protect the blade when it is not in use. They are constructed of titanium and are autoclavable or suitable for chemical sterilization.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.