Vannas Spring Scissors | Avantor
Vannas Spring Scissors
Vannas Spring Scissors
Certificates
Vannas Spring Scissors
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These Vannas scissors are specialized spring scissors designed for precision cutting and often used by scientific researchers.
  • Straight blades
  • 0.015×0.015 mm super fine tips
  • 3 mm blades
  • Stainless steel
  • Autoclavable
Compared with other spring scissors, Vannas scissors are distinguished by their delicate, thin, sharp tips. The slender, pointed tips allows for precise cutting with minimal damage to surrounding tissues. Manufactured with surgical grade stainless steel, these scissors are durable and resistant to corrosion. As with all your surgical instruments, these scissors should be cleaned immediately after use to ensure a long service life and minimize staining or corrosion.
Product Family Options
Informations sur le produit
  • Désignation
    Longueur
    Matière
    Availability
    Price
  • Vannas scissors, straight tip
    8 cm (3")
    Stainless steel
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $683.70
    $683.70
    Each
       $683.70
    Supplier Number: 501778
  • Vannas scissors, curved tip
    8 cm (3")
    Stainless steel
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $717.75
    $717.75
    Each
       $717.75
    Supplier Number: 501839