ContactUsLinkComponent
Bone Wax
Catalogue # 77662-834
New Product
Bone Wax
Catalogue # 77662-834
Supplier Number:  501771
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Bone wax
  • Size:
    2.5 g
  • Cat. no.:
    77662-834
Spécifications
À propos de cet article
Achieve homeostasis of bone by applying a small amount of sterile bone wax. The wax is a combination of beeswax, isopropyl palmitate and mineral oil.
  • Sterile
  • Stops bleeding at the site