Olsen-Hegar Needle Holders with Suture Scissors
Olsen-Hegar Needle Holders with Suture Scissors
Olsen-Hegar Needle Holders with Suture Scissors
Certificates
Olsen-Hegar Needle Holders with Suture Scissors
5 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Olsen-Hegar needle holders are a dual purpose surgical instrument, combining the functions of a needle holder and surgical scissors into one instrument.
  • Combine needle holders and surgical scissors in one surgical instrument
  • Straight
  • Serrated jaws, scissors combination
  • Tungsten carbide jaws
  • Surgical grade stainless steel or premium German steel
This is ideal for suturing, because you can grasp the needle for stitching and use the integrated scissors for trimming the ends without reaching for a different surgical instrument. Streamline your suturing process and save time. Simultaneously, you can reduce the risk of dropping needles.
The Olsen-Hegar needle holders are often used in orthopedics, plastic surgery, and any application requiring surturing. Life science researchers doing in vivo animal studies like implanting electrodes would also find this instrument useful. Researchers working with microinjection or microfluidics would also benefit from the dual use of this needle holder/scissors combination.
The Olsen-Hegar needle holders with suture Scissors are ring handled and have a locking mechanism. The lock is important when the surgical tool is not in use. Clamping the jaws tightly closed minimizes the chances of injury and contamination. Keeping the instrument securely closed also helps to reduce wear and tear on the instrument and keeps the tips properly aligned.
These Olsen-Hegar needle holders have straight, serrated jaws. They all have tungsten carbide inserts, which are more durable and improve the life of the needle holders. They are manufactured from surgical grade stainless steel or premium German steel, both of which are suitable for sterilization by autoclave or chemical sterilant. Always clean your surgical instruments promptly after use. That will keep them performing well.
Product Family Options
Informations sur le produit
  • Matière
    Longueur
    Availability
    Price
  • Stainless steel
    12 cm (4.75")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $406.31
    $406.31
    Each
       $406.31
    Supplier Number: 501312
  • Stainless steel
    14 cm (5.5")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $316.32
    $316.32
    Each
       $316.32
    Supplier Number: 500227
  • Stainless steel
    16.5 cm (6.5")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $316.32
    $316.32
    Each
       $316.32
    Supplier Number: 501725
  • German steel
    14 cm (5.5")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $248.08
    $248.08
    Each
       $248.08
    Supplier Number: 500227-G
  • German steel
    16.5 cm (6.5")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $333.38
    $333.38
    Each
       $333.38
    Supplier Number: 501725-G