Senn Retractor | Avantor
Senn Retractor
Senn Retractor
Certificates
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The Senn Retractor is a hand retractor which lets to gently hold back or retract tissues for better visibility and access to the surgical site.
  • Flat, thin, and curved blades
  • Sharp or blunt tips
  • Double ended
  • Fenestrated and solid blades
  • Surgical grade stainless steel which is autoclavable
This is commonly used in procedures involving skin, subcutaneous tissues, or superficial muscles. The retractor's blades can be inserted between layers of tissue and gently maneuvered to create the desired exposure. Use it for wound exposure, soft tissue dissection, or orthopedic applications.
The two ended handle has both fenestrated and solid blades which are both flat, thin, and curved to facilitate tissue retraction and maneuverability.
Product Family Options
Informations sur le produit
  • Longueur
    Matière
    Tip Configuration
    Availability
    Price
  • 15.9 cm (6.25")
    Stainless steel
    Shart tip
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $92.46
    $92.46
    Each
       $92.46
    Supplier Number: 501327
  • 15.9 cm (6.25")
    Stainless steel
    Blunt tip
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $133.83
    $133.83
    Each
       $133.83
    Supplier Number: 501718