KIMAX® Reservoir Buret, Automatic Self-Zeroing, Serialized | Avantor
KIMAX® Reservoir Buret, Automatic Self-Zeroing, Serialized
KIMAX® Reservoir Buret, Automatic Self-Zeroing, Serialized
Certificates
KIMAX® Reservoir Buret, Automatic Self-Zeroing, Serialized
KIMAX® Reservoir Buret, Automatic Self-Zeroing, Serialized
4 Options Available Below
Product Details & Documents
Class-A burettes ideal for repeat titrations requiring traceable volumetric accuracy.
  • Self-zeroing
  • Easy-to-read durable black enamel scale
  • Permanently marked with an individual serial number and traceable to NIST standards
  • Manufactured from 33 expansion, low extractable borosilicate glass
Filling and the overflow tubes accept 1/4" I.D. flexible tubing.
Certifications: Designed from ASTM E287, Class A requirements. Manufactured to the specifications found in ASTM E694. Calibrated to the accuracy requirements found in ASTM E542.
Delivery information: Supplied with a Certificate of Graduation Accuracy.
Product Family Options
Informations sur le produit
  • Capacité
    Longueur
    Intervalle de graduation
    Plage de graduation
    Tolerance
    Availability
    Price
  • 10 ml
    520 mm
    0.05 ml
    0.5-10 mL
    ±0.02 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $590.75
    $590.75
    Prix promotionnel pour les nouveaux clients
    Case of 1
       $590.75
    Supplier Number: 17054F-10
  • 25 ml
    580 mm
    0.10 ml
    1.0-25 mL
    ±0.03 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $591.60
    $591.60
    Prix promotionnel pour les nouveaux clients
    Case of 1
       $591.60
    Supplier Number: 17054F-25
  • 50 ml
    740 mm
    0.10 ml
    1.5-50 mL
    ±0.05 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $611.15
    $611.15
    Prix promotionnel pour les nouveaux clients
    Case of 1
       $611.15
    Supplier Number: 17054F-50
  • 100 ml
    770 mm
    0.20 ml
    3.0-100 mL
    ±0.10 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $760.76
    $760.76
    Prix promotionnel pour les nouveaux clients
    Case of 1
       $760.76
    Supplier Number: 17054F-100