À propos de cet article
Mini blumenthal rongeurs are used for removing bone or other hard tissue during surgical procedures, often in dental procedures. Blumenthal rongeurs are designed with a pointed, triangular cutting tip for precise cutting and shaping of bone and other hard tissue.
- Slight angle
- 2 mm jaw width
- Double spring handle
These rongeurs have double spring handles, giving you more control. They can be used in a variety of procedures, including orthopedic surgeries, neurosurgery, and plastic surgery. They may be used in studies examining bone growth or regeneration, or in studies exploring the effects of various drugs or treatments on bone tissue.
The mini blumenthal rongeurs are smaller than the standard version, and it is for use in more restricted spaces. These surgical rongeurs are available in surgical grade stainless steel or premium German steel. Sterilize them with a chemical sterilant or by autoclaving. To properly care for your surgical instruments, they should be cleaned promptly after every use.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.