ContactUsLinkComponent
VWR® Signature™ High-Performance Horizontal Air Flow Ovens
Fournisseur:  VWR International
Certificats
undefined
VWR® Signature™ High-Performance Horizontal Air Flow Ovens
Fournisseur:  VWR International
Spécifications
  • Catalog No:
  • 52202-258
  • 52202-509
  • Temperature range:
  • 15 °C above ambient to 260 °C
  • 15 °C above ambient to 260 °C
Spécifications
Informations connexes
Services: VWR CATALYST™ offers validation services including IQ, OQ, and PQ protocol development and execution, as well as asset management and calibration services (US only); call 888-793-2300 for more details.
White Glove Installation and Verification Service–the white glove installation includes unpacking, inspection, removal/disposal of packing materials, transporting order via elevator and merchandise set up in the room of your choice, installation of casters or leveling feet, power and electrical connections inspected, supply voltage verified, functional and performance verification documented. Our engineers will move the unit from the customer's receiving dock to the lab within the same building. This service does NOT include transportation from vendor warehouse to customer's receiving dock.
Frequently Bought Together