Absorbent Paper Points | Avantor
Absorbent Paper Points
Absorbent Paper Points
Certificates
Absorbent Paper Points
4 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Absorbent paper points are commonly used in laboratories, particularly in the field of dentistry and endodontics. These paper points are small, sterile, and highly absorbent.
  • Made from higly absorbent paper
  • Uniformly rolled to ensure correct size
  • Good for small volume absorption in confined areas
These sterile paper points are tapered to target specific areas and soak up small volumes of liquid. A variety of color-coded sizes are available, including extra fine, #15 fine, medium and coarse.
Packaging: Package of 100 to 200, depending on the size of the paper points.
Product Family Options
Informations sur le produit
  • Color
    Type
    Packaging
    Availability
    Price
  • Green
    C - Coarse
    Pack of 100
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $63.28
    $63.28
    Each
       $63.28
    Supplier Number: 504179
  • White
    Extra fine
    Pack of 200
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $63.28
    $63.28
    Each
       $63.28
    Supplier Number: 504180
  • White
    Fine
    Pack of 200
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $63.28
    $63.28
    Each
       $63.28
    Supplier Number: 504181
  • Blue
    Medium
    Pack of 200
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $63.28
    $63.28
    Each
       $63.28
    Supplier Number: 504182