Micro Scissors, Diamond Coated Blades | Avantor
Micro Scissors, Diamond Coated Blades
Micro Scissors, Diamond Coated Blades
Certificates
Micro Scissors, Diamond Coated Blades
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The special diamond coated blades will increase the longevity of the cutting edge and improve the qulaity of the cut.
Used to reduce wear on razor blades and industrial metal cutting tools, this coating is now being used in biomedical applications. Some studies have shown that even after double the normal usage of the cutting implement, no blade wear was detected.
Product Family Options
Informations sur le produit
  • Désignation
    Longueur
    Matière
    Blade Length
    Curvature
    Availability
    Price
  • Micro scissors, sharp, DLC coated
    12 cm
    Stainless steel
    8 mm
    Curved
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $768.86
    $768.86
    Each
       $768.86
    Supplier Number: 503364
  • Micro scissors, sharp, DLC coated
    12 cm
    Stainless steel
    8 mm
    Straight
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $768.86
    $768.86
    Each
       $768.86
    Supplier Number: 503365