Wilmer Iris Scissors
Wilmer Iris Scissors
Iris scissor, 45° angled, serrated, 10.5 cm (4"), Stainless steel
Catalog # 77663-192
Supplier #:  503248
Wilmer Iris Scissors
Catalog # 77663-192
Wilmer Iris Scissors
Iris scissor, 45° angled, serrated, 10.5 cm (4"), Stainless steel
Price
$175.19
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Iris scissors were originally designed for ophthalmic procedures but are now used in a wide range of applications. The tips are sharp, and they come in curved, straight or angled styles.
Specifications
  • Désignation:
    Iris scissor, 45° angled, serrated
  • Longueur:
    10.5 cm (4")
  • Matière:
    Stainless steel