Westcott Student Spring Scissors | Avantor
Westcott Student Spring Scissors
Westcott Student Spring Scissors
Certificates
Westcott Student Spring Scissors
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Westcott student spring scissors are popular spring handled scissors that automatically reopen after each cut. Westcott brand spring scissors give you an ergonomic and comfortable grip, reducing hand strain and fatigue during repetitive cutting tasks.
  • Slightly curved tip gives you greater visibility at the incision site
Westcott spring scissors are perfect for ambidextrous use, often preferred by left-handed individuals. The tips of these surgical scissors are slightly curved, and they are available with blunt or sharp tips. These Westcott scissors are perfect for student use. They are constructed of surgical grade stainless steel and are suitable for autoclaving or chemical sterilization.
Product Family Options
Informations sur le produit
  • Désignation
    Longueur
    Matière
    Blade Type
    Curvature
    Availability
    Price
  • Wescott student scissors
    11 cm (4.3")
    Stainless steel
    Blunt
    Slightly curved
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $274.95
    $274.95
    Each
       $274.95
    Supplier Number: 503222
  • Wescott student scissors
    11 cm (4.3")
    Stainless steel
    Sharp
    Slightly curved
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $274.95
    $274.95
    Each
       $274.95
    Supplier Number: 503223