VWR® 750 Benchtop Dissolved Oxygen Meters
Fournisseur: SCHULER SCIENTIFIC
New Product
Certificats
À propos de cet article
The VWR-DO750 meter provide accurate and reliable measurements of dissolved oxygen concentration and dissolved oxygen saturation.
- High resolution LCD display (5.7")
- Support for air-saturated water or zero oxygen calibration
- IP54 rating
- Automatic or manual temperature compensation capabilities
- Auto-read, timed-read, and continuous-read measurement options
- Data capacity of 500 sets (GLP-compliant)
- Auto barometric pressure compensation
- Manual salinity factor correction
- Three-year meter warranty
The easy to use VWR-DO750 benchtop dissolved oxygen meter provides accurate dissolved oxygen measurements for water quality testing applications. The large, backlit LCD display allows results to easily be viewed during operation.
Ordering information: Connect the meter easily to the VWR® dot matrix printer (10799-132; sold separately) to print measurement reports.
For application specific recommendations, please reference 'VWR® Electrode Selection Guide' located in the related information section.
For application specific recommendations, please reference 'VWR® Electrode Selection Guide' located in the related information section.
Delivery: All meters include in-use cover and electrode stand.
Spécifications
- Catalog No:
- 77618-934
- 77618-932
- Model:
- VWR-DO750A
- VWR-DO750