Vannas Spring Scissors | Avantor
Vannas Spring Scissors
Vannas Spring Scissors
Certificates
Vannas Spring Scissors
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Our Vannas spring scissors are specialized spring scissors designed for precision cutting and often used by scientific researchers.
  • Perfect for ambidextrous use, prefered by left hand users
  • 500 µm extra fine tips
  • Angled, straight or curved tips
  • Titanium or surgical grade stainless steel
Compared with other spring scissors, Vannas spring scissors are distinguished by their delicate, thin, sharp tips. The slender, pointed tips allows for precise cutting with minimal damage to surrounding tissues. Like all spring scissors, they are perfect for ambidextrous use and are often preferred by left-handed people. Manufactured with surgical grade stainless steel, these scissors are durable and resistant to corrosion.
Our titanium scissors are lightweight and non-magnetic. These Vannas have a fine 500 µm tip and 8 mm blades, making them suitable for surgery on small animals or researchers working with delicate tissues. They are autoclavable. Like all fine surgical instruments, these Vannas surgical scissors should be cleaned promptly after every use.
Product Family Options
Informations sur le produit
  • Désignation
    Longueur
    Matière
    Blade Length
    Blade Type
    Availability
    Price
  • Vannas spring scissors
    9 cm
    Stainless steel
    8 mm
    Straight
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $218.99
    $218.99
    Each
       $218.99
    Supplier Number: 501777
  • Vannas spring scissors
    9 cm
    Stainless steel
    8 mm
    Curved
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $218.99
    $218.99
    Each
       $218.99
    Supplier Number: 501930