Magnetize/Demagnetize Instrument | Avantor
Magnetize/Demagnetize Instrument
Demagnetize/magnetize surgical instruments, 2×2×1"
Catalog # 77662-944
Fournisseur #:  501900
Magnetize/Demagnetize Instrument
Catalog # 77662-944
Magnetize/Demagnetize Instrument
Demagnetize/magnetize surgical instruments, 2×2×1"
Price
$116.83
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Instantly magnetize or demagnetize small surgical instruments and small steel parts that are over 1.3" in length and 0.125" in diameter.
  • No batteries or electricity required
  • Box is high quality molded ABS plastic, impact resistant and cadmium free
  • The innovative step design lets you control the level of magnetism
To Magnetise a tool: Insert tool into the "+" opening and slide back and forth. Continue this procedure until the desired level of magnetism is reached.
To Demagnetise a tool: Insert tool at the lowest "−" opening. Slide back and forth. Continue the procedure, moving up to the next step until the tool is completely demagnetised.
Spécifications
  • Désignation:
    Demagnetize/magnetize surgical instruments, 2×2×1"