ContactUsLinkComponent
TenSette® Single Channel Pipettes, Variable Volume
TenSette® Single Channel Pipettes, Variable Volume
Catalog # 104085-678
Supplier:  Hach
undefined
TenSette® Single Channel Pipettes, Variable Volume
Catalog # 104085-678
Supplier:  Hach
Supplier Number:  1970010
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Volume:
    1.0 - 10 ml
  • Cat. no.:
    104085-678
  • Increments:
    1 ml
Specifications
About this item
With these pipettes users are able to perform routine laboratory pipetting quickly, economically, and accurately. The pipettes dispense from 0.1 to 1 ml or from 1 to 10 ml in 0.1 or 1.0 ml increments.
  • Save time, reduces glassware cleanup and assures accuracy
  • Specifically designed for use with Hach Voluette and PourRite standards in Standard Additions accuracy checks
Ordering information: Set includes pipette, manual and 50 disposable tips.