Test Kits, Drop Count Tests
: Hach
The drop count test is a simple, easy and cost-effective way to test for many parameters like chlorine, chloride, dissolved oxygen, carbon dioxide, sulphite, nitrogen, formaldehyde, and more.
- Portable - ideal for use in field or in lab
- Used as a quick reference or for confirmatory test with color indicator
- Reagents supplied with the kit
: Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Frequently Bought Together