VWR® M750 Benchtop pH/Ion/Conductivity/Dissolved Oxygen Meters
Supplier: VWR International
New Product
Certificates
About this item
The VWR-M750 benchtop multi-parameter meter provides accurate and reliable measurements of pH, conductivity, ion concentration, and dissolved oxygen.
- High resolution LCD display (5.7")
- Five-point calibration for pH with selectable pH buffer group
- Built-in ion methods including F⁻, NH⁴⁺ and NA⁺
- IP54 rating
- Automatic or manual temperature compensation capabilities
- Auto-read, timed-read, and continuous-read measurement options
- Three-point conductivity calibration with standard recognition
- Dissolved oxygen calibration supports air-sautrated water or zero oxygen
- Ability to save 500 data sets for each parameter (GLP-compliant)
The easy to use VWR-M750 benchtop meter provides the ability to perform various measurement parameters such as pH, TDS, ORP and dissolved oxygen with a single meter. The large, backlit LCD display allows results to easily be viewed during operation.
Ordering information: Three-year meter warranty. Connect the meter easily to the VWR® dot matrix printer (10799-132; sold separately) to print measurement reports. For application specific recommendations, please reference 'VWR® Electrode selection guide' located in the related information section.
Frequently Bought Together