Replacement Tips for Reusable Rapid Biopsy Punch
New Product
Stainless steel replacement tips for use with reusable rapid biopsy punch.
- Replacement tips for reusable punch
- Designed for heavy, numerous and precise sampling operations up to 2000 punches
- Tip composition: Stainless steel 440C with heat treatment HRC 58 hardness
- For use with reusable rapid punch biopsy kit
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together