Reflex Clips System | Avantor
Reflex Clips System
Reflex Clips System
Certificates
Reflex Clips System
5 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
For smal animal wound closure.
  • Clips are non stertile
  • Appliers are non sterile but can be autoclaved
Clips are made up of stainless steel material.
Packaging: Clips are packed 100 per box.
Caution: This item is designed for research only, not intended for human use.
Product Family Options
Clips
Informations sur le produit
  • Désignation
    Availability
    Price
  • Reflex clips, 7 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $184.92
    $184.92
    Pack of 100
       $184.92
    Supplier Number: 500344
  • Reflex clips, 9 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $167.89
    $167.89
    Pack of 100
       $167.89
    Supplier Number: 500346
Appliers
Informations sur le produit
  • Désignation
    Availability
    Price
  • Reflex clip applier for 7 mm clips
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $525.56
    $525.56
    Each
       $525.56
    Supplier Number: 500343
  • Reflex clip applier for 9 mm clips
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $525.56
    $525.56
    Each
       $525.56
    Supplier Number: 500345
Clip removing forcep
Informations sur le produit
  • Désignation
    Availability
    Price
  • Reflex clip removing forcep of 7 and 9 mm clips, 12 cm long
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $389.30
    $389.30
    Each
       $389.30
    Supplier Number: 500347