Microelectrode Holders, MPH6S
Microelectrode Holders, MPH6S
Microelectrode Holders, MPH6S
Microelectrode holder-half-cells couple fluid-filled glass micropipettes to high input impedance amplifiers. A Ag/AgCl pellet (or a silver wire) molded into the holder body provides stable potential. Electrical connection is made via male 2 mm pins or female 2 mm sockets.
  • Female luer
  • Screw cap
The pipette may be mounted axially or at right angles to the holder. Pipettes are held with screw-caps or rubber gaskets (without caps). Filling WPI microelectrode holders with electrolytes containing chloride results in stable electrode potential. Suitable electrolytes include KCl, NaCl and CaCl₂. Holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. The holder style you select will depend on your experimental application, space, and instrumentation.
Product Family Options
  • For Use With
    Désignation
  • 1.0 mm standard single capillary tubing
    Microelectrode holder (MPH6S)
    Each
    Each - $131.14
    $131.14
    Each
       $131.14
     : MPH6S10
  • 1.2 mm standard single capillary tubing
    Microelectrode holder (MPH6S)
    Each
    Each - $143.80
    $143.80
    Each
       $143.80
     : MPH6S12
  • 1.5 mm standard single capillary tubing
    Microelectrode holder (MPH6S)
    Each
    Each - $143.78
    $143.78
    Each
       $143.78
     : MPH6S15
  • 2.0 mm standard single capillary tubing
    Microelectrode holder (MPH6S)
    Each
    Each - $143.78
    $143.78
    Each
       $143.78
     : MPH6S20