Steam Sterilization Indicator Labels | Avantor
Steam Sterilization Indicator Labels
Labels, Roll of 5000
Catalog # 19872-046
Supplier:   Crosstex
Fournisseur #:  NMCPI-S01
Steam Sterilization Indicator Labels
Catalog # 19872-046
Supplier:  Crosstex
Steam Sterilization Indicator Labels
Steam Sterilization Indicator Labels
Labels, Roll of 5000
Price
$414.36
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Self-adhesive, nontoxic, chemical process indicators change from blue to pink upon exposure to 10 minutes of saturated steam at 121 °C (250 °F) or 2 minutes at 134 °C (273 °F).
Longer exposure will result in a brighter shade of pink. The blue to pink color transition is sensitive to time, temperature, and the presence of saturated steam. Water-based ink is made without the use of heavy metals. Indicators are stable for at least 18 months from date of manufacture when they are stored at room temperature.
Certifications: Conform to AAMI ST60 and ISO 11140-1, Section 6 for Class 1 Process Indicators.
Documents
Spécifications
  • Désignation:
    Labels, Roll of 5000