To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Description:Coomassie Plus (Bradford) Reagent
- Size:300 mL
- Cat. no.:CAPI23238
- No. of Assays/Kit:200 standard assays or 1000 microplate assays
- Supplier No.:23238
The Bradford protein assay is the preferred colorimetric assay for quantifying total protein concentration
- Sensitive, fast and consistent protein assay kit
- Determine protein concentrations from 1 to 1500 μg/mL
- Convenient microplate or cuvette format
- Compatible with reducing sugars, reducing substances and thiols
This modification of the well-known Bradford method greatly reduces the tendency of Coomassie reagents to give non linear response curves by a formulation that substantially improves linearity for a defined range of protein concentration. In addition, Coomassie Plus Reagent results in significantly less protein-to-protein variation than is observed with other Bradford-type Coomassie formulations.
The Pierce™ Coomassie Plus Protein Assay is a ready-to-use, reducing agent-compatible, improved Bradford assay reagent to quickly measure (A595 nm) total protein concentration compared to a protein standard.